Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290541.1 | NP_001277470.1 | 1 - 2 | 189 | 68 | |
NM_001290543.1 | NP_001277472.1 | 1 - 2 | 189 | 68 | ||
NM_139306.3 | NP_647467.1 | 1 - 2 | 189 | 68 | ||
GenBank mRNA | AF282864.1 | - | 1 - 2 | 148 | 68 | |
AK085306.1 | - | 1 - 2 | 185 | 68 | ||
AY312515.1 | - | 1 - 2 | 114 | 68 | ||
BC059819.1 | - | 1 - 2 | 189 | 6839525 | 68 | |
CB723138.1 | - | 1 - 2 | 189 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |