Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130194.1 | NP_001123666.1 | 4 - 5 | 566 | 75 | |
NR_052011.1 | 4 - 5 | 807 | 75 | |||
GenBank mRNA | AY450428.1 | - | 4 - 5 | 808 | 75 | |
BC019528.1 | - | 3 - 4 | 540 | 4208860 | 75 | |
BC031186.1 | - | 3 - 4 | 527 | 4989959 | 75 | |
BC036157.1 | - | 4 - 5 | 566 | 4982413 | 75 | |
BC036163.1 | - | 4 - 5 | 607 | 4984410 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |