Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285988.1 | NP_001272917.1 | 5 - 6 | 708 | 116 | |
NM_001285989.1 | NP_001272918.1 | 5 - 6 | 708 | 116 | ||
NM_145436.2 | NP_663411.2 | 5 - 6 | 708 | 116 | ||
GenBank mRNA | AK039377.1 | - | 5 - 6 | 505 | 116 | |
AK134425.1 | - | 4 - 5 | 420 | 116 | ||
AK148278.1 | - | 5 - 6 | 564 | 116 | ||
AK161471.1 | - | 5 - 6 | 577 | 116 | ||
BC023187.1 | - | 5 - 6 | 566 | 116 | ||
BC157955.1 | - | 5 - 6 | 707 | 9088128 | 116 | |
BC172100.1 | - | 5 - 6 | 707 | 9054386 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |