Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145554.2 | NP_663529.2 | 2 - 3 | 338 | 63 | |
GenBank mRNA | AK029008.1 | - | 2 - 3 | 303 | 63 | |
AK148340.1 | - | 2 - 3 | 300 | 63 | ||
AK159800.1 | - | 2 - 3 | 282 | 63 | ||
AK167290.1 | - | 2 - 3 | 287 | 63 | ||
BC021467.1 | - | 2 - 3 | 287 | 5067154 | 63 | |
BC066808.1 | - | 2 - 3 | 281 | 30055031 | 63 | |
BC067411.1 | - | 2 - 3 | 281 | 30092248 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |