Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144896.4 | NP_659145.1 | 4 - 5 | 654 | 90 | |
GenBank mRNA | AK028193.1 | - | 4 - 5 | 653 | 90 | |
AK034702.1 | - | 4 - 5 | 655 | 90 | ||
AK053978.1 | - | 4 - 5 | 657 | 90 | ||
AK134831.1 | - | 4 - 5 | 516 | 90 | ||
AK166231.1 | - | 4 - 5 | 665 | 90 | ||
BC005709.1 | - | 4 - 5 | 656 | 3157973 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |