Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042655.1 | NP_001036120.1 | 15 - 16 | 1864 | 63 | |
GenBank mRNA | AK039566.1 | - | 15 - 16 | 1679 | 63 | |
AK046883.1 | - | 14 - 15 | 1801 | 63 | ||
AK081318.1 | - | 13 - 14 | 1858 | 63 | ||
AK083153.1 | - | 15 - 16 | 1682 | 63 | ||
AK087754.1 | - | 1690 | 63 | |||
AK136705.1 | - | 15 - 16 | 1689 | 63 | ||
AK149917.1 | - | 15 - 16 | 1676 | 63 | ||
AK153732.1 | - | 15 - 16 | 1676 | 63 | ||
AK170821.1 | - | 15 - 16 | 1676 | 63 | ||
BC017607.1 | - | 15 - 16 | 1881 | 4507589 | 63 | |
BC035449.1 | - | 15 - 16 | 1663 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |