Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146062.3 | NP_666174.2 | 6 - 7 | 737 | 73 | |
NM_175363.4 | NP_780572.1 | 5 - 6 | 530 | 73 | ||
GenBank mRNA | AK081606.1 | - | 5 - 6 | 523 | 73 | |
AK150904.1 | - | 5 - 6 | 481 | 73 | ||
AK160138.1 | - | 5 - 6 | 503 | 73 | ||
BC031486.1 | - | 6 - 7 | 690 | 3157250 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |