Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146107.2 | NP_666219.1 | 8 - 9 | 998 | 88 | |
GenBank mRNA | AK078508.1 | - | 8 - 9 | 1015 | 88 | |
AK088839.1 | - | 8 - 9 | 1031 | 88 | ||
BC023020.1 | - | 8 - 9 | 998 | 5375425 | 88 | |
BC024861.1 | - | 3 - 4 | 322 | 88 | ||
BC027134.1 | - | 1664 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |