Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011763.2 | NP_035893.1 | 2 - 3 | 188 | 73 | |
GenBank mRNA | AK007823.1 | - | 2 - 3 | 167 | 73 | |
AK029052.1 | - | 2 - 3 | 174 | 73 | ||
BC059273.1 | - | 1 - 2 | 52 | 2647280 | 73 | |
BY132524.1 | - | 2 - 3 | 173 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |