Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164609.1 | NP_001158081.1 | 2 - 3 | 159 | 83 | |
NM_001164610.1 | NP_001158082.1 | 2 - 3 | 159 | 83 | ||
NM_001164611.1 | NP_001158083.1 | 2 - 3 | 159 | 83 | ||
NM_029945.3 | NP_084221.2 | 2 - 3 | 159 | 83 | ||
GenBank mRNA | AK019466.2 | - | 2 - 3 | 160 | 83 | |
AK031685.1 | - | 2 - 3 | 158 | 83 | ||
AK035892.1 | - | 2 - 3 | 151 | 83 | ||
AK036055.1 | - | 2 - 3 | 151 | 83 | ||
AK036380.1 | - | 2 - 3 | 155 | 83 | ||
AK129354.1 | - | 2 - 3 | 136 | 83 | ||
AK138720.1 | - | 2 - 3 | 133 | 83 | ||
AK164234.1 | - | 2 - 3 | 151 | 83 | ||
AK164456.1 | - | 2 - 3 | 133 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |