Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164609.1 | NP_001158081.1 | 3 - 4 | 254 | 83 | |
NM_001164610.1 | NP_001158082.1 | 3 - 4 | 254 | 83 | ||
NM_001164611.1 | NP_001158083.1 | 3 - 4 | 254 | 83 | ||
NM_029945.3 | NP_084221.2 | 3 - 4 | 254 | 83 | ||
GenBank mRNA | AK019466.2 | - | 3 - 4 | 255 | 83 | |
AK031029.1 | - | 3 - 4 | 248 | 83 | ||
AK031685.1 | - | 3 - 4 | 253 | 83 | ||
AK035892.1 | - | 3 - 4 | 246 | 83 | ||
AK036055.1 | - | 3 - 4 | 246 | 83 | ||
AK036380.1 | - | 3 - 4 | 250 | 83 | ||
AK129354.1 | - | 3 - 4 | 231 | 83 | ||
AK138720.1 | - | 3 - 4 | 228 | 83 | ||
AK164234.1 | - | 3 - 4 | 246 | 83 | ||
AK164456.1 | - | 3 - 4 | 228 | 83 | ||
BC026767.1 | - | 3 - 4 | 227 | 5137008 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |