Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dner | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152915.1 | NP_690879.1 | 7 - 8 | 1751 | 71 | |
GenBank mRNA | AB067650.1 | - | 7 - 8 | 1400 | 71 | |
AF370126.1 | - | 7 - 8 | 1407 | 71 | ||
AK044597.1 | - | 7 - 8 | 1514 | 71 | ||
AK152755.1 | - | 2 - 3 | 154 | 71 | ||
AK153235.1 | - | 2 - 3 | 154 | 71 | ||
AY032924.1 | - | 7 - 8 | 1407 | 71 | ||
BC022636.1 | - | 2 - 3 | 214 | 71 | ||
BC034634.1 | - | 7 - 8 | 1751 | 5365190 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |