Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pddc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172116.4 | NP_742114.3 | 1 - 2 | 122 | 90 | |
GenBank mRNA | AK081187.1 | - | 1 - 2 | 82 | 90 | |
AK084273.1 | - | 1 - 2 | 78 | 90 | ||
AK140975.1 | - | 1 - 2 | 73 | 90 | ||
AK150323.1 | - | 1 - 2 | 76 | 90 | ||
AK150354.1 | - | 1 - 2 | 76 | 90 | ||
AK151259.1 | - | 1 - 2 | 76 | 90 | ||
AK151794.1 | - | 1 - 2 | 76 | 90 | ||
AK151959.1 | - | 1 - 2 | 76 | 90 | ||
AK153110.1 | - | 1 - 2 | 76 | 90 | ||
AK161205.1 | - | 1 - 2 | 88 | 90 | ||
BC023835.1 | - | 1 - 2 | 92 | 5345186 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |