Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Limd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172397.3 | NP_765985.1 | 3 - 4 | 227 | 77 | |
GenBank mRNA | AK012581.1 | - | 3 - 4 | 201 | 77 | |
AK032430.1 | - | 3 - 4 | 228 | 77 | ||
AK040486.1 | - | 3 - 4 | 228 | 77 | ||
AK049809.1 | - | 3 - 4 | 197 | 77 | ||
AK167349.1 | - | 3 - 4 | 197 | 77 | ||
AK169829.1 | - | 3 - 4 | 198 | 77 | ||
AK171794.1 | - | 3 - 4 | 506 | 77 | ||
BC068130.1 | - | 3 - 4 | 171 | 6413206 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |