Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138677.2 | NP_619618.1 | 8 - 9 | 1596 | 120 | |
GenBank mRNA | AB042828.1 | - | 8 - 9 | 1585 | 120 | |
AK030943.1 | - | 8 - 9 | 1597 | 120 | ||
AK037889.1 | - | 8 - 9 | 1159 | 120 | ||
AK082920.1 | - | 8 - 9 | 1549 | 120 | ||
AK129090.1 | - | 8 - 9 | 1563 | 120 | ||
AK135969.1 | - | 3 - 4 | 460 | 120 | ||
AK152724.1 | - | 8 - 9 | 1572 | 120 | ||
AK152979.1 | - | 8 - 9 | 1568 | 120 | ||
BC023237.1 | - | 8 - 9 | 1544 | 2654113 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |