Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800022590.mm.1 |
NM_001190786.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022590.mm.1 |
NM_001190786.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022590.mm.1 |
NM_001190800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022590.mm.1 |
NM_001190800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022590.mm.1 |
NM_172284.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022590.mm.1 |
NM_172284.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022590.mm.1 |
TC0800002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800022586.mm.1 |
NM_001190786.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022586.mm.1 |
NM_001190786.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022586.mm.1 |
NM_001190800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022586.mm.1 |
NM_001190800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022586.mm.1 |
NM_172284.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022586.mm.1 |
NM_172284.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022586.mm.1 |
TC0800002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800022591.mm.1 |
NM_001190786.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022591.mm.1 |
NM_001190786.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022591.mm.1 |
NM_001190800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022591.mm.1 |
NM_001190800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022591.mm.1 |
NM_172284.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022591.mm.1 |
NM_172284.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022591.mm.1 |
TC0800002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800022589.mm.1 |
NM_001190786.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022589.mm.1 |
NM_001190786.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022589.mm.1 |
NM_001190800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022589.mm.1 |
NM_001190800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022589.mm.1 |
NM_172284.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022589.mm.1 |
NM_172284.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022589.mm.1 |
TC0800002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800022593.mm.1 |
NM_001190786.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022593.mm.1 |
NM_001190786.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022593.mm.1 |
NM_001190800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022593.mm.1 |
NM_001190800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022593.mm.1 |
NM_172284.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022593.mm.1 |
NM_172284.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022593.mm.1 |
TC0800002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800022588.mm.1 |
NM_001190786.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022588.mm.1 |
NM_001190786.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022588.mm.1 |
NM_001190800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022588.mm.1 |
NM_001190800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022588.mm.1 |
NM_172284.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022588.mm.1 |
NM_172284.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022588.mm.1 |
TC0800002935.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800022592.mm.1 |
NM_001190786.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022592.mm.1 |
NM_001190786.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022592.mm.1 |
NM_001190800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022592.mm.1 |
NM_001190800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022592.mm.1 |
NM_172284.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022592.mm.1 |
NM_172284.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800022592.mm.1 |
TC0800002935.mm.1 |
MTA TC |