Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177843.1 | NP_001171314.1 | 12 - 13 | 1412 | 60 | |
NM_001177844.1 | NP_001171315.1 | 1 - 2 | 540 | 60 | ||
NM_172475.3 | NP_766063.3 | 12 - 13 | 1042 | 60 | ||
GenBank mRNA | AK029073.1 | - | 1 - 2 | 519 | 60 | |
AK077327.1 | - | 1 - 2 | 541 | 60 | ||
AK082365.1 | - | 12 - 13 | 988 | 60 | ||
AK148414.1 | - | 1 - 2 | 539 | 60 | ||
AK171532.1 | - | 1 - 2 | 468 | 60 | ||
AK196650.1 | - | 1 - 2 | 61 | |||
BC058672.1 | - | 12 - 13 | 1033 | 6830456 | 60 | |
BC082808.1 | - | 12 - 13 | 1419 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |