Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100024636.mm.1 |
NM_001114662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024636.mm.1 |
NM_001114662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024636.mm.1 |
NM_172510.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024636.mm.1 |
NM_172510.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024636.mm.1 |
TC0100003068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024631.mm.1 |
NM_001114662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024631.mm.1 |
NM_001114662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024631.mm.1 |
NM_172510.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024631.mm.1 |
NM_172510.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024631.mm.1 |
TC0100003068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024632.mm.1 |
NM_001114662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024632.mm.1 |
NM_001114662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024632.mm.1 |
NM_172510.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024632.mm.1 |
NM_172510.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024632.mm.1 |
TC0100003068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024638.mm.1 |
NM_001114662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024638.mm.1 |
NM_001114662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024638.mm.1 |
NM_172510.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024638.mm.1 |
NM_172510.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024638.mm.1 |
TC0100003068.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100024633.mm.1 |
NM_001114662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024633.mm.1 |
NM_001114662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024633.mm.1 |
NM_172510.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024633.mm.1 |
NM_172510.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100024633.mm.1 |
TC0100003068.mm.1 |
MTA TC |