Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172614.3 | NP_766202.3 | 6 - 7 | 853 | 77 | |
GenBank mRNA | AK031524.1 | - | 5 - 6 | 3305 | 77 | |
AK045675.1 | - | 7 - 8 | 916 | 77 | ||
AK048655.1 | - | 6 - 7 | 854 | 77 | ||
AK079084.1 | - | 1 - 2 | 179 | 77 | ||
AK082652.1 | - | 6 - 7 | 2088 | 77 | ||
BC059216.1 | - | 6 - 7 | 1191 | 77 | ||
BC080307.1 | - | 7 - 8 | 749 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |