Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172704.3 | NP_766292.2 | 5 - 6 | 544 | 87 | |
GenBank mRNA | AK053156.1 | - | 5 - 6 | 526 | 87 | |
AK089983.1 | - | 5 - 6 | 536 | 87 | ||
AK171781.1 | - | 5 - 6 | 525 | 87 | ||
AK172542.1 | - | 5 - 6 | 525 | 87 | ||
BC079852.1 | - | 5 - 6 | 514 | 87 | ||
BC085257.1 | - | 5 - 6 | 518 | 5414963 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |