Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172707.3 | NP_766295.2 | 2 - 3 | 566 | 118 | |
GenBank mRNA | AK004686.2 | - | 2 - 3 | 277 | 118 | |
AK088893.1 | - | 2 - 3 | 206 | 118 | ||
AK147112.1 | - | 2 - 3 | 369 | 118 | ||
AK166168.1 | - | 2 - 3 | 484 | 118 | ||
AK166416.1 | - | 2 - 3 | 570 | 118 | ||
AK166623.1 | - | 2 - 3 | 469 | 118 | ||
AK168141.1 | - | 2 - 3 | 463 | 118 | ||
AK168512.1 | - | 2 - 3 | 212 | 118 | ||
AK169379.1 | - | 2 - 3 | 315 | 118 | ||
AK171283.1 | - | 2 - 3 | 435 | 118 | ||
AK198041.1 | - | 1 - 2 | 141 | 118 | ||
AK202060.1 | - | 1 - 2 | 141 | 118 | ||
AK214320.1 | - | 1 - 2 | 142 | 118 | ||
BC046832.1 | - | 2 - 3 | 267 | 6812937 | 118 | |
M27073.1 | - | 2 - 3 | 275 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |