Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172833.2 | NP_766421.1 | 12 - 13 | 1768 | 134 | |
GenBank mRNA | AK041919.1 | - | 12 - 13 | 1773 | 134 | |
AK085071.1 | - | 12 - 13 | 1768 | 134 | ||
BC110487.1 | - | 13 - 14 | 1775 | 40048907 | 134 | |
BC110488.1 | - | 12 - 13 | 1742 | 40048908 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protease Mammalian Gene Collection Mus musculus Probe spans exons |