Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglecg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172900.3 | NP_766488.2 | 9 - 10 | 1746 | 105 | |
GenBank mRNA | AK036698.1 | - | 6 - 7 | 3202 | 105 | |
AK042488.1 | - | 9 - 10 | 1747 | 105 | ||
AK156766.1 | - | 5 - 6 | 3155 | 105 | ||
AY210400.1 | - | 8 - 9 | 1601 | 105 | ||
EF419770.1 | - | 8 - 9 | 1609 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |