Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172964.4 | NP_766552.3 | 10 - 11 | 1401 | 63 | |
GenBank mRNA | AK087523.1 | - | 10 - 11 | 1414 | 63 | |
AK141901.1 | - | 10 - 11 | 1351 | 63 | ||
AK162051.1 | - | 4 - 5 | 441 | 63 | ||
BC066788.1 | - | 10 - 11 | 1391 | 30049271 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |