Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163847.1 | NP_001157319.1 | 6 - 7 | 1442 | 61 | |
NM_001163848.1 | NP_001157320.1 | 6 - 7 | 1528 | 61 | ||
NM_001163849.1 | NP_001157321.1 | 6 - 7 | 1442 | 61 | ||
NM_001163850.1 | NP_001157322.1 | 5 - 6 | 1424 | 61 | ||
NM_001163851.1 | NP_001157323.1 | 5 - 6 | 1510 | 61 | ||
NM_001163852.1 | NP_001157324.1 | 6 - 7 | 1660 | 61 | ||
NM_001163853.1 | NP_001157325.1 | 5 - 6 | 1488 | 61 | ||
NM_173186.4 | NP_775278.3 | 5 - 6 | 1510 | 61 | ||
GenBank mRNA | AK036088.1 | - | 6 - 7 | 1395 | 61 | |
AK045194.1 | - | 5 - 6 | 1392 | 61 | ||
AK049306.1 | - | 5 - 6 | 1489 | 61 | ||
AK049754.1 | - | 5 - 6 | 1398 | 61 | ||
AK083833.1 | - | 6 - 7 | 1414 | 61 | ||
AK084693.1 | - | 5 - 6 | 1464 | 61 | ||
AK086293.1 | - | 5 - 6 | 1469 | 61 | ||
AK129304.1 | - | 5 - 6 | 1442 | 61 | ||
AK138431.1 | - | 6 - 7 | 1403 | 61 | ||
AK147647.1 | - | 6 - 7 | 1528 | 61 | ||
AK161054.1 | - | 6 - 7 | 1662 | 61 | ||
BC080845.1 | - | 3 - 4 | 1275 | 6850962 | 61 | |
BC094417.1 | - | 4 - 5 | 1344 | 30943242 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |