Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173788.3 | NP_776149.1 | 19 - 20 | 3137 | 57 | |
GenBank mRNA | AK158195.1 | - | 14 - 15 | 1688 | 57 | |
BC023420.1 | - | 6 - 7 | 778 | 57 | ||
BC037458.1 | - | 14 - 15 | 1593 | 57 | ||
BC042470.1 | - | 19 - 20 | 3137 | 5137261 | 57 | |
BC049896.1 | - | 9 - 10 | 1232 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |