Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174850.3 | NP_777275.2 | 13 - 14 | 2897 | 105 | |
GenBank mRNA | AB182579.1 | - | 13 - 14 | 2748 | 105 | |
AK044547.1 | - | 13 - 14 | 2749 | 105 | ||
AK151086.1 | - | 13 - 14 | 2896 | 105 | ||
AK164043.1 | - | 13 - 14 | 2868 | 105 | ||
AK165558.1 | - | 13 - 14 | 2747 | 105 | ||
AK220308.1 | - | 13 - 14 | 2674 | 105 | ||
BC043300.1 | - | 13 - 14 | 2711 | 5025620 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |