Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253813.1 | NP_001240742.1 | 6 - 7 | 836 | 108 | |
NM_175344.4 | NP_780553.2 | 5 - 6 | 773 | 108 | ||
GenBank mRNA | AK089300.1 | - | 5 - 6 | 684 | 108 | |
AK159411.1 | - | 6 - 7 | 787 | 108 | ||
BC060732.1 | - | 5 - 6 | 773 | 6409332 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |