Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175481.5 | NP_780690.2 | 13 - 14 | 1681 | 82 | |
GenBank mRNA | AK032029.1 | - | 13 - 14 | 1682 | 82 | |
AK080582.1 | - | 13 - 14 | 1810 | 82 | ||
BC118010.2 | - | 12 - 13 | 1596 | 40096690 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |