Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prtg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175485.4 | NP_780694.3 | 12 - 13 | 2306 | 116 | |
GenBank mRNA | AK083540.1 | - | 10 - 11 | 1742 | 116 | |
AY630257.1 | - | 12 - 13 | 2376 | 116 | ||
BC116336.1 | - | 10 - 11 | 1739 | 116 | ||
BC116337.1 | - | 10 - 11 | 1739 | 116 | ||
DQ360114.1 | - | 12 - 13 | 2306 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |