Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290625.1 | NP_001277554.1 | 5 - 6 | 817 | 75 | |
NM_001290626.1 | NP_001277555.1 | 4 - 5 | 815 | 75 | ||
NM_177345.4 | NP_796319.1 | 5 - 6 | 1143 | 75 | ||
GenBank mRNA | AK042154.1 | - | 4 - 5 | 805 | 75 | |
AK052045.1 | - | 5 - 6 | 811 | 75 | ||
AK132263.1 | - | 5 - 6 | 807 | 75 | ||
AK148428.1 | - | 5 - 6 | 1132 | 75 | ||
AK191438.1 | - | 1 - 2 | 137 | 75 | ||
BC043296.1 | - | 5 - 6 | 1042 | 75 | ||
BC090644.1 | - | 4 - 5 | 717 | 30629263 | 75 | |
BC096618.1 | - | 5 - 6 | 1116 | 6392317 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |