Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177407.4 | NP_803126.1 | 1 - 2 | 203 | 69 | |
GenBank mRNA | AK083245.1 | - | 1 - 2 | 204 | 69 | |
AK122410.2 | - | 1 - 2 | 199 | 69 | ||
AK163179.1 | - | 1 - 2 | 204 | 69 | ||
BC031745.1 | - | 1 - 2 | 184 | 4506005 | 69 | |
X14836.1 | - | 1 - 2 | 187 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |