Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gale | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178389.3 | NP_848476.1 | 2 - 3 | 408 | 61 | |
GenBank mRNA | AK009083.1 | - | 3 - 4 | 275 | 61 | |
AK146184.1 | - | 3 - 4 | 300 | 61 | ||
BC027438.1 | - | 2 - 3 | 408 | 4224021 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |