Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030180.2 | NP_084456.2 | 9 - 10 | 1716 | 89 | |
GenBank mRNA | AK036864.1 | - | 8 - 9 | 1105 | 89 | |
AK047620.1 | - | 9 - 10 | 1716 | 89 | ||
CF741928.1 | - | 1 - 2 | 107 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |