Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177772.4 | NP_808440.2 | 7 - 8 | 732 | 81 | |
GenBank mRNA | AK028764.1 | - | 7 - 8 | 729 | 81 | |
AK132518.1 | - | 7 - 8 | 768 | 81 | ||
BC112376.1 | - | 7 - 8 | 720 | 40057331 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |