Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252458.1 | NP_001239387.1 | 6 - 7 | 1167 | 83 | |
NM_178900.4 | NP_849231.1 | 5 - 6 | 1452 | 83 | ||
GenBank mRNA | AK037030.1 | - | 5 - 6 | 1453 | 83 | |
AK041207.1 | - | 4 - 5 | 672 | 83 | ||
AK170761.1 | - | 6 - 7 | 1131 | 83 | ||
BC095949.1 | - | 6 - 7 | 1117 | 4950957 | 83 | |
BC096444.1 | - | 6 - 7 | 1135 | 4216827 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |