Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkx2-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008700.2 | NP_032726.1 | 1 - 2 | 538 | 117 | |
GenBank mRNA | AK142146.1 | - | 1 - 2 | 443 | 117 | |
BC139299.1 | - | 1 - 2 | 384 | 8862321 | 117 | |
BC139303.1 | - | 1 - 2 | 384 | 8862325 | 117 | |
L20300.1 | - | 1 - 2 | 355 | 117 | ||
X75415.1 | - | 1 - 2 | 603 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |