Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025811.3 | NP_080087.1 | 9 - 10 | 2122 | 130 | |
GenBank mRNA | AK002650.1 | - | 9 - 10 | 1707 | 130 | |
AK004564.1 | - | 9 - 10 | 1769 | 130 | ||
AK033384.1 | - | 9 - 10 | 2123 | 130 | ||
BC041104.1 | - | 7 - 8 | 1091 | 130 | ||
BC050211.1 | - | 7 - 8 | 977 | 130 | ||
BC095956.1 | - | 9 - 10 | 2066 | 5330002 | 130 | |
BC138190.1 | - | 9 - 10 | 1959 | 8861209 | 130 | |
BC138191.1 | - | 9 - 10 | 1959 | 8861210 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |