Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153527.2 | NP_705755.2 | 6 - 7 | 774 | 70 | |
GenBank mRNA | AF419292.2 | - | 6 - 7 | 919 | 70 | |
AK005984.1 | - | 6 - 7 | 791 | 70 | ||
AK139680.1 | - | 747 | 70 | |||
BC048501.1 | - | 6 - 7 | 774 | 6705915 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |