Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwf19l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027545.2 | NP_081821.1 | 9 - 10 | 1519 | 105 | |
GenBank mRNA | AK033457.1 | - | 2 - 3 | 263 | 105 | |
AK037905.1 | - | 1 - 2 | 581 | 105 | ||
AK045423.1 | - | 9 - 10 | 1520 | 105 | ||
AK083386.1 | - | 9 - 10 | 1532 | 105 | ||
AK167727.1 | - | 2 - 3 | 242 | 105 | ||
BC043480.1 | - | 9 - 10 | 1486 | 105 | ||
BC125483.1 | - | 9 - 10 | 1556 | 40129998 | 105 | |
BC125485.1 | - | 9 - 10 | 1556 | 40130000 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |