Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zufsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028287.2 | NP_082563.1 | 2 - 3 | 201 | 70 | |
GenBank mRNA | AK012263.1 | - | 2 - 3 | 212 | 70 | |
AK046191.1 | - | 2 - 3 | 204 | 70 | ||
AK052071.1 | - | 2 - 3 | 185 | 70 | ||
AK077304.1 | - | 2 - 3 | 249 | 70 | ||
AK077730.1 | - | 2 - 3 | 222 | 70 | ||
AK160928.1 | - | 2 - 3 | 200 | 70 | ||
BC067065.1 | - | 1 - 2 | 115 | 70 | ||
BC089514.1 | - | 2 - 3 | 214 | 30252639 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |