Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081978.2 | NP_001075447.1 | 1 - 2 | 58 | 72 | |
NM_001290371.1 | NP_001277300.1 | 1 - 2 | 58 | 72 | ||
NM_009666.4 | NP_033796.1 | 1 - 2 | 58 | 72 | ||
GenBank mRNA | AK029358.1 | - | 1 - 2 | 62 | 72 | |
CD773419.1 | - | 1 - 2 | 62 | 72 | ||
D31768.1 | - | 1 - 2 | 49 | 72 | ||
D31769.1 | - | 1 - 2 | 49 | 72 | ||
KC731591.1 | - | 1 - 2 | 58 | 72 | ||
S74899.1 | - | 1 - 2 | 58 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |