Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177995.1 | NP_001171466.1 | 10 - 11 | 2807 | 59 | |
NM_001291026.1 | NP_001277955.1 | 10 - 11 | 2801 | 59 | ||
NM_001291029.1 | NP_001277958.1 | 10 - 11 | 2810 | 59 | ||
NM_027504.3 | NP_081780.3 | 10 - 11 | 2804 | 59 | ||
GenBank mRNA | AB078338.1 | - | 10 - 11 | 2696 | 59 | |
AK083829.1 | - | 10 - 11 | 2692 | 59 | ||
AK173230.1 | - | 2 - 3 | 986 | 59 | ||
BC059838.1 | - | 10 - 11 | 2788 | 6409778 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |