Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029546.2 | NP_083822.1 | 3 - 4 | 286 | 64 | |
GenBank mRNA | AB041855.1 | - | 3 - 4 | 247 | 64 | |
AK088254.1 | - | 3 - 4 | 287 | 64 | ||
AK088983.1 | - | 3 - 4 | 287 | 64 | ||
BC112325.1 | - | 3 - 4 | 252 | 5355810 | 64 | |
BC117903.1 | - | 3 - 4 | 267 | 64 | ||
BC128480.1 | - | 3 - 4 | 267 | 40097069 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |