Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pof1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181579.1 | NP_853557.1 | 4 - 5 | 559 | 141 | |
GenBank mRNA | AF309775.1 | - | 3 - 4 | 452 | 141 | |
AK036922.1 | - | 4 - 5 | 564 | 141 | ||
AK086511.1 | - | 4 - 5 | 560 | 141 | ||
AK137653.1 | - | 4 - 5 | 560 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |