Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198100.2 | NP_932768.2 | 1 - 2 | 350 | 63 | |
GenBank mRNA | AK014173.1 | - | 2 - 3 | 312 | 63 | |
BC057590.1 | - | 1 - 2 | 350 | 6810986 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |