Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D10Jhu81e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138601.2 | NP_613067.1 | 1 - 2 | 177 | 91 | |
GenBank mRNA | AK003866.1 | - | 1 - 2 | 178 | 91 | |
BC013475.1 | - | 1 - 2 | 170 | 4237589 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |