Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175638.3 | NP_783569.1 | 1 - 2 | 711 | 126 | |
GenBank mRNA | AY184228.1 | - | 1 - 2 | 680 | 126 | |
AY187027.1 | - | 1 - 2 | 636 | 126 | ||
AY260059.1 | - | 1 - 2 | 674 | 126 | ||
BC037114.1 | - | 1 - 2 | 721 | 126 | ||
BC043677.1 | - | 1 - 2 | 84 | 126 | ||
BC096453.1 | - | 1 - 2 | 667 | 4234311 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |