Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144836.2 | NP_659085.1 | 7 - 8 | 1180 | 89 | |
GenBank mRNA | AK040717.1 | - | 7 - 8 | 1181 | 89 | |
AK050184.1 | - | 7 - 8 | 1163 | 89 | ||
AK050277.1 | - | 7 - 8 | 1057 | 89 | ||
BC018306.1 | - | 6 - 7 | 818 | 4193755 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |