Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013657.5 | NP_038685.3 | 16 - 17 | 1900 | 99 | |
GenBank mRNA | AK034239.1 | - | 16 - 17 | 1959 | 99 | |
AK077231.1 | - | 5 - 6 | 1023 | 99 | ||
AK140348.1 | - | 2436 | 99 | |||
BC066852.1 | - | 16 - 17 | 1900 | 30093230 | 99 | |
X85994.1 | - | 16 - 17 | 1852 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |